Learn More
Description
- Sterile
- BIO-CERT™ CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, pyrogens, and non-cytotoxic
- Smooth-walled
- cellGrade™ plus surface treatment
- Available in PC or PET membrane materials for different optical requirements and cell visibility
- Multiple membrane pore sizes for different applications
- Packaged in 12 strips of four wells per pack
Applications:
Secretion studies, co-culture, migration studies, chemotaxis assays
Specifications
Specifications
| Bottom Shape | Flat |
| Capacity (Metric) | 400 μL |
| Color | Clear |
| Culture Area | 0.6 cm2 |
| Material | Polystyrene |
| Sterility | Sterile |
| Surface Treatment | cellGrade plus |
| Type | Cell Culture Insert Strip |
| Well Count | 4 |
| Pore Size | 1 μm |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.