Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ BRANDplates™ Smooth-Walled Insert Strips

Catalog No. 14380849 Shop All BrandTech Scientific Products
Change view
Click to view available options
Pore Size:
0.4 μm
1 μm
3 μm
8 μm
Supportive Membrane:
PC
PET
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Supportive Membrane
14-380-849 3 μm PC
14-380-833 0.4 μm PC
14-380-857 8 μm PC
14-380-841 1 μm PC
14-380-861 8 μm PET
14-380-837 0.4 μm PET
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-380-849 Supplier BRANDTECH™ BRAND™ Supplier No. 782840
Add to Cart
Add to Cart

Available with either polycarbonate (PC) or polyethylene terephthalate (PET) membranes.

  • Sterile
  • BIO-CERT™ CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, pyrogens, and non-cytotoxic
  • Smooth-walled
  • cellGrade™ plus surface treatment
  • Available in PC or PET membrane materials for different optical requirements and cell visibility
  • Multiple membrane pore sizes for different applications
  • Packaged in 12 strips of four wells per pack

Applications:

Secretion studies, co-culture, migration studies, chemotaxis assays

TRUSTED_SUSTAINABILITY

Specifications

Bottom Shape Flat
Capacity (Metric) 400 μL
Color Clear
Culture Area 0.6 cm2
Material Polystyrene
Sterility Sterile
Surface Treatment cellGrade plus
Type Cell Culture Insert Strip
Well Count 4
Pore Size 3 μm
Quantity 12/Pk.
Supportive Membrane PC
Height (Metric) 13.4 mm
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.