Learn More
BRANDTECH™ BRAND™ Valves and Seals for seripettor™ and seripettor™ pro
Catalog No. 0384096 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Seripettor bottletop dispenser
Catalog No. For Use With (Equipment)
03-840-96 Seripettor bottletop dispenser
1 options
Catalog No. 03-840-96 Supplier BRANDTECH™ BRAND™ Supplier No. 6790
Add to Cart
Add to Cart
Replacement valves and seals for use with seripettor™ bottletop dispensers.
Filling valve set
  • Includes: 1 x filling valve body, 1 x filling valve, 1 x discharge valve, and 2 x sealing rings
Sealing rings
  • Five per pack
  • Also compatible with the Quiksip™ bottletop aspirator
TRUSTED_SUSTAINABILITY
Specifications
Product Type Valve Set
Autoclavable Autoclavable
For Use With (Application) Dispensing liquid
For Use With (Equipment) Seripettor bottletop dispenser
Includes Filling valve, discharge valve, and two seals
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov