Learn More
BRANDTECH™ BRAND™ PCR Cap Tool
Catalog No. 14388172 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
No. per Pack:
1 Ea.
Catalog No. No. per Pack
14-388-172 1 Ea.
1 options
Catalog No. 14-388-172 Supplier BRANDTECH™ BRAND™ Supplier No. 781419
Add to Cart
Edge
Add to Cart
For use with attaching and detaching caps on a PCR Plate.
  • Polyamide construction
  • Attaches and removes caps on PCR plates
TRUSTED_SUSTAINABILITY
Specifications
Compatibility All Plates Sealed with Strip Caps
For Use With (Application) Attaching and removing caps on a PCR plate
For Use With (Equipment) PCR plate caps
Material Polyamide
No. per Pack 1 Ea.
Product Type PCR Cap Tool
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov