Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Deep Well Plates

Catalog No. 14380804 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Volume (Metric) Well:
0.3 mL
0.5 mL
1.1 mL
2.2 mL
Format:
Standard
Well Shape:
Round
Round Bottom (U-well)
Square, Conical Bottom (V-well)
Square, Round Bottom (U-well)
Profile:
Low Profile
U-bottom
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Well Format Well Shape Profile
14-380-804 0.3 mL Standard Square, Conical Bottom (V-well) U-bottom
13-882-50 0.5 mL Standard Round Bottom (U-well) U-bottom
13-882-51 1.1 mL Standard Round Bottom (U-well) U-bottom
13-882-52 1.1 mL Standard Round U-bottom
13-882-53 2.2 mL Standard Square, Round Bottom (U-well) U-bottom
14-380-803 1.1 mL Standard Round Bottom (U-well) Low Profile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-380-804 Supplier BRANDTECH™ BRAND™ Supplier No. 701355
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Seamlessly integrates into multi-channel systems and automation workflows.

BRAND™ Deep Well Plates are designed for a broad range of applications, including sample storage, high-throughput screening (HTS), cell and tissue culture, serial dilutions, and reagent transfers. Most plates are manufactured from high-quality virgin polypropylene, with one highly transparent polystyrene variant for applications requiring optical clarity.

  • Available in 96- and 384-well formats
  • Manufactured from high-quality virgin polypropylene or polystyrene
  • BIO-CERT™ PCR Quality are free from human DNA, DNase, RNase, and pyrogens
  • Polypropylene plates are autoclavable, chemically resistant, and suitable for storage down to -80°C (-112°F)
  • Polystyrene plate offers excellent transparency and is suitable for storage down to -20°C (-4°F)
  • ANSI/SLAS 1 and 4 compliant for compatibility with automated systems
  • Alphanumeric well labeling and stackable design for efficient organization
  • Notched corners ensure correct orientation in robotic and automated liquid-handling systems
  • Compatible with sealing mats or adhesive films to minimize evaporation and contamination
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Format Standard
Array 16 x 24
Autoclavable Autoclavable
Color Clear
Product Type Deep-well Plate
Material Polypropylene
Volume (Metric) Well 0.3 mL
No. of Wells 384
Sterility Non-sterile
Surface Treatment Untreated
Well Shape Square, Conical Bottom (V-well)
For Use With (Application) Flexible mat covers or adhesive sealing films to reduce sample evaporation and contamination
No. per Pack 48
Profile U-bottom
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.