Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Non-Sterile Microcentrifuge Tubes

Catalog No. 14380820 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Color:
Amber
Blue
Clear
Green
Orange
Yellow
Quantity:
10,000/Pk.
500/Pk.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
14-380-820 Amber 500/Pk.
13-882-100 Clear 500/Pk.
14-380-815 Clear 10,000/Pk.
14-380-816 Yellow 500/Pk.
14-380-817 Blue 500/Pk.
14-380-818 Green 500/Pk.
14-380-819 Orange 500/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 14-380-820 Supplier BRANDTECH™ BRAND™ Supplier No. 780525

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

Fits most common rotors and may be spun up to 30,000 RCF. Non-Sterile Microcentrifuge Tubes are highly translucent and free from flash, air bubbles, and streaks. Polypropylene flat snap caps form a tight, leak-free, easy-open seal. Tubes have a uniform wall thickness for consistent, repeatable results.

Fits most common rotors and may be spun up to 30,000 RCF (g) (10,000 x g for 0.5mL; 20,000 x g for 1.5mL; 30,000 x g for 1.5mL and 2.0mL with lid closure at 20°C for 20 min).
  • Manufactured from virgin polypropylene
  • Highly translucent tubes are free from flash, air bubbles, and streaks.
  • Caps form a tight, leak-free seal that is easy to open, even when wearing gloves, and are flat with consistent thickness for trouble-free piercing.
  • Tubes have uniform wall thickness for consistent, repeatable results.
  • Subdivisions for approximate volume determination.
  • Autoclavable at 120°C (2bar), according to DIN EN 285.
  • Lid closure are certified endotoxin, DNA-, DNase-, and RNase-free.
  • Colored tubes simplify sample identification
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Endotoxin, RNase-, ATP- and DNA-free
Color Amber
Volume (Metric) 1.5 mL
Quantity 500/Pk.
Closure Material Polypropylene
For Use With (Equipment) Centrifuge
Sterility Non-sterile
Material Polypropylene
Max. Force 30,000 x g
Closure Type Flat Snap Cap
Autoclavable Autoclavable
Format Tube
Product Type Microcentrifuge Tube
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.