Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Self-adhesive Plate Sealing Film

Catalog No. 14380811 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Material:
Aluminum
Polyester
Polyethylene, Polypropylene
Polypropylene
Rayon
Vinyl
For Use With (Application):
Automation
Cell and tissue culture
Cold storage
ELISA, PCR
ELISA, Real-time PCR
Luminescence measurements
Storage
Storage, fluorescence measurements
No. per Pack:
100 Sheets
50 Sheets
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material For Use With (Application) No. per Pack
14-380-811 Vinyl Luminescence measurements 50 Sheets
13-883-09 Polyester ELISA, PCR 100 Sheets
13-882-95 Rayon Cell and tissue culture 100 Sheets
14-380-807 Polypropylene Storage 100 Sheets
13-882-329 Aluminum Cold storage 100 Sheets
13-878-201 Rayon Cell and tissue culture 50 Sheets
14-380-809 Polyethylene, Polypropylene Automation 50 Sheets
14-380-810 Vinyl Storage, fluorescence measurements 50 Sheets
13-878-200 Polyester ELISA, Real-time PCR 100 Sheets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 14-380-811 Supplier BRANDTECH™ BRAND™ Supplier No. 701372
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Securely seals multiwell plates. Self-adhesive Plate Sealing Film can be applied directly to plates and removed without the use of expensive equipment, or can be applied with a hard rubber roller to ensure secure and uniform attachment of the film to the plate.

  • Easily apply to plates and remove without the use of expensive equipment.
  • Application with a hard rubber roller will ensure secure and uniform attachment of the film to the plate.
  • Sealing films are available in different materials especially suited for storage, immunoassay, or cell and tissue culture:
    • Polyethylene
    • Polyester
    • Aluminum
    • Rayon
    • Polypropylene
    • Vinyl
TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Vinyl
Sterility Non-sterile
Autoclavable Not Autoclavable
No. per Pack 50 Sheets
Evaporation Barrier No
For Use With (Application) Luminescence measurements
Pre-slit No
Product Type Sealing Film
Adhesive Pressure Sensitive
Thickness 14.1 mil.
Pierceable No
Temperature Range (Metric) -40°C to +80°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.