Learn More
BRANDTECH™ BRAND™ Dispensette™III, Dispensette™ Organic Remote Dispensing System Parts and Accessories
Shop All BRANDTECH Scientific ProductsDescription
- Dispensette™ parts and accessories allow the Dispensette™ to be mounted on a wall, a ring stand, or on lab furniture.
- Dispenses accurate volumes directly from drums and bulk refills.
- A filter in the drum adapter minimizes risk of contaminating high-purity reagents.
- A quick-release connector with integrated valves allows quick changing of the bulk container.
- The remote dispensing system allows storage of the drum up to 10 meters (30 feet) away from the Dispensette™.
- Maximum delivery height is approximately 1.2 m
Specifications
Specifications
| Product Type | Table/Shelf Clamp |
| For Use With (Equipment) | DispensetteIII, Dispensette Organic |
| Quantity | 1 Ea. |