Learn More
BRANDTECH™ VITLAB™ Snap-On Lid for Graduated HDPE Buckets
Catalog No. 14379258 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Material:
LDPE
Catalog No. Material
14-379-258 LDPE
1 options
Catalog No. 14-379-258 Supplier BRANDTECH™ VITLAB™ Supplier No. 96293
Add to Cart
Add to Cart
Secure-fitting and transparent lid made from LDPE.
  • Snap-on cover
  • Provides a tight seal for VITLAB™ HDPE buckets
  • Covers are made of transparent LDPE
  • Use part number 96293 for 5 L bucket
  • Use part number 96593 for 10 L bucket
TRUSTED_SUSTAINABILITY
Specifications
Product Type Lid, Bucket
Material LDPE
Autoclavable Not Autoclavable
WARNING: Reproductive Harm - www.P65Warnings.ca.gov