Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ VITLAB™ LDPE Bottles with Caps

Catalog No. 14379093 Shop All BrandTech Scientific Products
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
Closure Size:
GL 18
GL 25
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
14-379-093 250 mL GL 25
14-379-091 100 mL GL 18
14-379-094 500 mL GL 25
14-379-092 50 mL GL 18
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-379-093 Supplier BRANDTECH™ VITLAB™ Supplier No. 94989
Add to Cart
Add to Cart

VITLAB™ round bottles are made of low density polyethylene, with polypropylene screw caps

  • Light weight and break-resistant
TRUSTED_SUSTAINABILITY

Specifications

Capacity 8.45 oz., 250 mL
Capacity (Metric) 250 mL
Capacity (English) 8.45 oz.
Color Translucent Yellow
Diameter (English) 2.28 in.
Material LDPE
Quantity 12/Pk.
Sterility Non-sterile
Certifications/Compliance EC 10/2011
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 135 mm
Product Type Reagent Bottle
Height (English) 5.31 in.
Closure Material Polypropylene
Closure Size GL 25
Diameter (Metric) 58 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Autoclavable Not Autoclavable
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.