Learn More
BRANDTECH™ BRAND™ QuikSip™ Bottle Top Aspirator Accessory, Manifold
GSA_VA
Catalog No. 03840101 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
For Use With (Equipment):
QuikSip bottletop aspirator
Catalog No. For Use With (Equipment)
03-840-101 QuikSip bottletop aspirator
1 options
Catalog No. 03-840-101 Supplier BRANDTECH™ BRAND™ Supplier No. 704526
Add to Cart
Edge
Add to Cart
Autoclavable and used withQuikSip™ Bottle Top Aspirator
  • 8-channel
  • Polypropylene
  • Autoclavable (121°C)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Product Type 8 Channel Manifold
Autoclavable Autoclavable
Material Polypropylene
For Use With (Application) Removal of fluids from multiwell plates
For Use With (Equipment) QuikSip bottletop aspirator
WARNING: Reproductive Harm - www.P65Warnings.ca.gov