Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BrandTech™ BRAND™ PCR 8-Strip Clear Tubes

Catalog No. 13882304 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Tube strip; With attached domed strip cap; 8 x 0.2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
13-882-304 Tube strip; With attached domed strip cap; 8 x 0.2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-882-304 Supplier BrandTech™ BRAND™ Supplier No. 781330
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Manufactured from polypropylene without lubricants or chemical additives that can contaminate reagents and samples. PCR 8-Strip Tubes are RNase-, DNase-, and DNA-free and are compatible with most 24-, 32-, 96-, and 384-well thermal blocks.

Manufactured from high-quality virgin plastics for excellent, reproducible results in most PCR applications, and are manufactured without lubricants or chemical additives that can contaminate reagents and samples. Tube strips are available with or without caps and can be ordered separately.

  • Thin, uniform walls ensure consistent, precise thermal transfer for excellent results.
  • Autoclavable
  • RNase-, DNase-, and DNA-free.
  • Compatible with most 24-, 32-, 96-, and 384-well thermal blocks.
  • Available in convenient tube/cap sets.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Volume (Metric) 0.2 mL
Material Polypropylene
Dnase Free Yes
RNase Free Yes
Autoclavable Autoclavable
Closure Included Yes
Format Strip
Product Type Tube Strip
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.