Learn More
BrandTech™ BRAND™ PCR 8-Strip Clear Tubes
Catalog No. 13882305 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Strip tube/cap set; 8-strip; Clear
Catalog No.
13-882-305 Strip tube/cap set; 8-strip; Clear
1 options
Catalog No. 13-882-305 Supplier BrandTech™ BRAND™ Supplier No. 781326
Add to Cart
Add to Cart
Manufactured from polypropylene without lubricants or chemical additives that can contaminate reagents and samples. PCR 8-Strip Tubes are RNase-, DNase-, and DNA-free and are compatible with most 24-, 32-, 96-, and 384-well thermal blocks.
Manufactured from high-quality virgin plastics for excellent, reproducible results in most PCR applications, and are manufactured without lubricants or chemical additives that can contaminate reagents and samples. Tube strips are available with or without caps and can be ordered separately.
  • Thin, uniform walls ensure consistent, precise thermal transfer for excellent results.
  • Autoclavable
  • RNase-, DNase-, and DNA-free.
  • Compatible with most 24-, 32-, 96-, and 384-well thermal blocks.
  • Available in convenient tube/cap sets.
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Volume (Metric) 0.2 mL
Material Polypropylene
Dnase Free Yes
RNase Free Yes
Autoclavable Autoclavable
Closure Included Yes
Format Strip
Product Type Tubes and Flat Cap
WARNING: Reproductive Harm - www.P65Warnings.ca.gov