Learn More
BRANDTECH™ BRAND™ Polypropylene Thread Adapters for Bottle-Top Dispensers
Catalog No. 13688106 Shop All BrandTech Scientific Products
Change view
Click to view available options
Thread Size:
GL 33/28
GL 45/33
GL 45/38
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size
13-688-106 GL 33/28
13-688-321 GL 45/38
13-688-320 GL 45/33
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-688-106 Supplier BRANDTECH™ BRAND™ Supplier No. 704328
Only null left
Add to Cart
Edge
Add to Cart
Adapt current model Dispensette™ seripettor™, Titrette™ and QuikSip™ to common bottle sizes.
  • Polypropylene Construction
  • For use with Dispensette™ seripettor™ Titrette™ and QuikSip™ Bottle-top dispensers
TRUSTED_SUSTAINABILITY
Specifications
Product Type Bottle Thread Adapter
Material Polypropylene
For Use With (Equipment) Dispensette, seripettor, Titrette and QuikSip
Thread Size GL 33/28
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov