Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ VITLAB™ Reagent Bottles with Screw Caps

Catalog No. 14379102 Shop All BrandTech Scientific Products
Change view
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
2000 mL
250 mL
5000 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
14-379-102 10,000 mL
14-379-096 100 mL
14-379-351 1000 mL
14-379-100 2000 mL
14-379-352 5000 mL
14-379-097 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-379-102 Supplier BRANDTECH™ VITLAB™ Supplier No. 100989
Add to Cart
Add to Cart

VITLAB™ polypropylene reagent bottles with polypropylene screw caps have good chemical resistance. Autoclavable at 121 C for 20 minutes.

  • Good chemical resistance for long-term storage of liquids.
  • Screw caps help protect against outside contamination and leakage.
  • Autoclavable at 121 C, according to DIN EN 285
TRUSTED_SUSTAINABILITY

Specifications

Capacity 338.1 oz., 10000mL
Capacity (Metric) 10,000 mL
Capacity (English) 338.1 oz.
Color Transparent
Diameter (English) 8.74 in.
Material Polypropylene
Quantity 6/Pk.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Threaded
Height (Metric) 394 mm
Product Type Reagent Bottle
Height (English) 15.51 in.
Closure Material Polypropylene
Closure Size GL 63
Diameter (Metric) 222 mm
Closure Type Screw Cap
Autoclavable Not Autoclavable
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.