Learn More
BRANDTECH™ BRAND™ Replacement Parts for Dispensette™ Organic Bottle Top Dispensers: Discharge Valve Washer
Shop All BRANDTECH Scientific ProductsDescription
- Does not fit other dispensers
- Five each of filling washers, and discharge/SafetyPrime washers.
Specifications
Specifications
| Product Type | Discharge Tube Seal |
| Material | PTFE |
| For Use With (Equipment) | Dispensette III, organic and digital buret III |
| Includes | Five filing valve rings and five discharge/SafetyPrime seals |