Learn More
BRANDTECH™ BRAND™ Recirculation Tube for SafetyPrime Recirculation Valve
GSA_VA
Catalog No. 1387935 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Dispensette III, organic and HF bottletop dispenser with SafetyPrime venting valve
Catalog No. For Use With (Equipment)
13-879-35 Dispensette III, organic and HF bottletop dispenser with SafetyPrime venting valve
1 options
Catalog No. 13-879-35 Supplier BRANDTECH™ BRAND™ Supplier No. 8317
Add to Cart
Add to Cart
For Dispensette III, Digital Burette III and Titrette
  • Recirculation tube to fit BRAND Dispensette III, Dispensette Organic, Digital Buret III and Titrette instruments
  • Does not fit BRAND Dispensette S instruments
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Recirculation Tube
For Use With (Equipment) Dispensette III, organic and HF bottletop dispenser with SafetyPrime venting valve
WARNING: Reproductive Harm - www.P65Warnings.ca.gov