Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Transferpette™ -8/-12 Electronic Multichannel Pipette

Catalog No. 01671249 Shop All BrandTech Scientific Products
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
1 to 20 μL
10 to 200 μL
15 to 300 μL
5 to 100 μL
50 to 1250 μL
Number of Channels:
8
12
Increments:
0.02 μL
0.2 μL
1 μL
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Number of Channels Increments
01-671-249 50 to 1250 μL 8 1 μL
13-668-422 0.5 to 10 μL 8 0.02 μL
13-668-423 0.5 to 10 μL 12 0.02 μL
13-668-424 1 to 20 μL 8 0.02 μL
13-668-425 1 to 20 μL 12 0.02 μL
13-668-426 5 to 100 μL 8 0.2 μL
13-668-427 5 to 100 μL 12 0.2 μL
13-668-428 10 to 200 μL 8 0.2 μL
13-668-429 10 to 200 μL 12 0.2 μL
13-668-430 15 to 300 μL 8 0.2 μL
13-668-431 15 to 300 μL 12 0.2 μL
01-671-248 50 to 1250 μL 12 1 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 01-671-249 Supplier BRANDTECH™ BRAND™ Supplier No. 705398
Only null left
Add to Cart
Add to Cart

Includes

Battery, 120V AC adapter with charging cable, silicone oil, operating manual, pipette stand, reagent reservoir, tip cone tool, set of replacement V-rings

Versatile pipette offering five operating modes to improve productivity and increase sample throughput.

  • Choice of 8 or 12 channel models
  • Clear, legible LCD screen
  • Intuitive interface with user-friendly design allows easy operation
  • Soft touch pipetting key and adjustable finger rest provides added comfort
  • Rechargeable battery provides over 4000 pipetting cycles between charges
  • Multichannel V-rings on tip cones improves tip fit and reduces mounting and ejection force
  • Easily maintained

Operating modes

  • Pipetting (PIP Mode): previously set volume is aspirated into pipette tip and then discharged
  • Mixing of samples (PIPmix Mode): program for mixing liquids
  • Reverse Pipetting (revPIP Mode): program designed for greater accuracy
  • Pipetting for electrophoresis (GEL Mode): program for loading of electrophoresis gels
  • Dispensing (DISP Mode): single aspirated volume is dispensed in selected number of smaller, equal aliquots
TRUSTED_SUSTAINABILITY

Specifications

Accuracy ±12.5 μL
Battery Type Rechargeable
Compatible Tips 50 to 1250 μL Tips
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Gray
Disposable Reusable
For Use With Pipette Tip (1250 μL)
Grip Type Ergonomic
Material Plastic
Model DE-M
Number of Channels 8
Product Line Transferpette
Programmability Step-based
Sterility Non-sterile
Type Electronic Pipette
Volume (Metric) 50 to 1250 μL
Display LCD
Finger Style Ergonomic
Includes Battery Pack, Universal Power Adapter with Charging Cable, Shelf Mount, Silicone O-rings, Reagent Reservoir, Grease, Operating Manual
Increments 1 μL
Precision (0.25%) 3.125 μL
Warranty 1 Year
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.