Learn More
BRANDTECH™ BRAND™ Transferpette™ -8/-12 Electronic Multichannel Pipette
Shop All BrandTech Scientific ProductsDescription
Includes
Battery, 120V AC adapter with charging cable, silicone oil, operating manual, pipette stand, reagent reservoir, tip cone tool, set of replacement V-rings
- Choice of 8 or 12 channel models
- Clear, legible LCD screen
- Intuitive interface with user-friendly design allows easy operation
- Soft touch pipetting key and adjustable finger rest provides added comfort
- Rechargeable battery provides over 4000 pipetting cycles between charges
- Multichannel V-rings on tip cones improves tip fit and reduces mounting and ejection force
- Easily maintained
Operating modes
- Pipetting (PIP Mode): previously set volume is aspirated into pipette tip and then discharged
- Mixing of samples (PIPmix Mode): program for mixing liquids
- Reverse Pipetting (revPIP Mode): program designed for greater accuracy
- Pipetting for electrophoresis (GEL Mode): program for loading of electrophoresis gels
- Dispensing (DISP Mode): single aspirated volume is dispensed in selected number of smaller, equal aliquots
Specifications
Specifications
| Accuracy | ±0.2 μL |
| Battery Type | Rechargeable |
| Compatible Tips | 0.1 to 20 μL Nano-cap Tip, 0.5 to 20 μL Ultra Micro Tip, 0.5 to 10 μL Tip |
| Adjustable Tip Spacing | No |
| Autoclavable | Autoclavable |
| Color | Gray |
| Disposable | Reusable |
| For Use With | Pipette Tip (20 μL) |
| Grip Type | Ergonomic |
| Material | Plastic |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.