Learn More
BRANDTECH™ VACUUBRAND™ Rotary Vane Vacuum Pumps: Oil Mist Filters
Catalog No. 13889111 Shop All BrandTech Scientific Products
Change view
Click to view available options
Type:
Filter Element
Oil Mist Filter
For Use With:
RE/RZ 2, 2.5, 5, 6 Rotary Vane Pumps
RE/RZ 8, 9, 16 Rotary Vane Pumps
RZ 2.5, RZ 6 Oil Mist Filter
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With
13-889-111 Filter Element RZ 2.5, RZ 6 Oil Mist Filter
13-889-110 Oil Mist Filter RE/RZ 2, 2.5, 5, 6 Rotary Vane Pumps
13-889-114 Oil Mist Filter RE/RZ 8, 9, 16 Rotary Vane Pumps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-889-111 Supplier BRANDTECH™ VACUUBRAND™ Supplier No. 20698004
Add to Cart
Add to Cart
Oil Mist Filers and replacements for VACUUBRAND™ Rotary Vane Vacuum Pumps
  • For use with VACUUBRAND™ Rotary Vane Vacuum Pumps
  • Oil Mist Filters 'FO' trap up to 99% of oil mist generated by the pump, helping preserve the laboratory environment
TRUSTED_SUSTAINABILITY
Specifications
For Use With RZ 2.5, RZ 6 Oil Mist Filter
Type Filter Element
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov