Learn More
BRANDTECH™ VITLAB™ SAN Tall Form Graduated Cylinders, Class B
Catalog No. 14379167 Shop All BrandTech Scientific Products
Catalog No. 14-379-167 Supplier BRANDTECH™ VITLAB™ Supplier No. 65091
Add to Cart
Add to Cart
These Class B VITLAB™ SAN Tall Form Graduated Cylinders feature molded graduations with ring marks at major increments for excellent readability.
  • Made of crystal-clear Styrene Acrylonitrile (SAN).
  • Hexagonal base with bottom studs provides high stability and resists rolling.
  • Thermal stress up to 60°C does not cause tolerance limits to be permanently exceeded.
TRUSTED_SUSTAINABILITY
Specifications
Class Class B
Quantity 6 Pk.
Sterility Non-sterile
Base Type Hexagonal
Product Type Graduated Cylinder
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov