Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brass foil, alloy 260, 0.025mm (0.001in) thick, 7.2x45cm (3.0x18in)

Catalog No. AA47224KJ
Change view
Click to view available options
Quantity:
1 Ea.
5 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA47224KJ 5 Pk.
AA47224KG 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA47224KJ Supplier Thermo Scientific Chemicals Supplier No. 047224KJ

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Chemical Name or Material Brass foil
Name Note Alloy 260
Quantity 5 Pk.
Odor Odorless
Form Solid
MDL Number MFCD00171255

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.