Learn More
Diagnostic Biosystems BRCA1 BRCA1
Supplier:  Diagnostic Biosystems MOB42405
Encompass_Preferred
BRCA1 recognizes a 22kDa protein. This antibody stains 94% of ovarian carcinomas with a mutation in exon 11. However, staining was absent in 100% of tumors with mutations other than exon 11.
Catalog No. 50-238-5811
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.