Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam BRD-7389, p90 Ribosomal S6 Kinase Inhibitor, Solid, 25 mg
Description
- Potency: p90 ribosomal S6 kinase (RSK) inhibitor with IC50 values of 1.2 μM (RSK3), 1.5 μM (RSK1), and 2.4 μM (RSK2)
- Biological Activity: Upregulates insulin expression in pancreatic α-cells and demonstrates reprogramming effects in terminally differentiated cells
- Molecular Weight: 366.4 Da
- Composition: 100% 1-Phenethylamino-3H-naphtho[1,2,3-de]quinoline-2,7-dione
- Form: Solid
- Storage: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | p90 Ribosomal S6 Kinase (RSK) |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Form | Solid |
| Buffer | 100% 1-Phenethylamino-3H-naphtho[1,2,3-de]quinoline-2,7-dione |
| Content And Storage | Store at -20°C. |
| Formulation | 100% 1-Phenethylamino-3H-naphtho[1,2,3-de]quinoline-2,7-dione |
| Quantity | 25 mg |
| For Use With (Application) | p90 ribosomal S6 kinase inhibition |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?