Learn More
Abcam BRD-7389, p90 Ribosomal S6 Kinase Inhibitor, Solid, 5 mg
Description
- Molecular Weight: 366.4 Da
- Function: p90 ribosomal S6 kinase RSK inhibitor (IC50 values are 1.2, 1.5, and 2.4 μM for RSK3, RSK1, and RSK2 respectively)
- Biological Activity: Upregulates insulin expression in pancreatic α-cells; shows reprogramming effects in terminally differentiated cells
- Composition: 100% 1-Phenethylamino-3H-naphtho[1,2,3-de]quinoline-2,7-dione
- Form: Solid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | p90 Ribosomal S6 Kinase |
| Shipping Condition | Ambient |
| Buffer | 100% 1-Phenethylamino-3H-naphtho[1,2,3-de]quinoline-2,7-dione |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Formulation | 100% 1-Phenethylamino-3H-naphtho[1,2,3-de]quinoline-2,7-dione |
| Quantity | 5 mg |
| For Use With (Application) | p90 ribosomal S6 kinase RSK inhibition |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.