Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRD 7389, Tocris Bioscience™

Catalog No. 403750
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-375-0 50 mg
40-371-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 40-375-0 Supplier R&D Systems Supplier No. 4037/50
This item is not returnable. View return policy

Ribosomal S6 kinase inhibitor

Ribosomal S6 kinase (RSK) inhibitor (IC50 values are 1.2, 1.5 and 2.4 μM for RSK3, RSK1 and RSK2 respectively). Upregulates insulin expression in terminally differentiated pancreatic α-cells.

Chemical Identifiers

CAS 376382-11-5
Molecular Formula C24H18N2O2
Molecular Weight (g/mol) 366.42
InChI Key XASCINRGTHLHGM-UHFFFAOYSA-N
Synonym 1-2-phenylethyl amino-3h-naphtho 1,2,3-de quinoline-2,7-dione, 1-phenethylamino-3h-naphtho 1,2,3-de quinoline-2,7-dione, 16-2-phenylethyl amino-14-azatetracyclo 7.7.1.0 2 ,?.0 1 3 , 1 ? heptadeca-1 16 ,2 7 ,3,5,9 17 ,10,12-heptaene-8,15-dione, 16-2-phenylethyl amino-14-azatetracyclo 7.7.1.0 2 ,?.0 1 3 , 1 ? heptadeca-1 16 ,2 7 ,3,5,9,11,13 17-heptaene-8,15-dione, 16-2-phenylethyl amino-14-azatetracyclo 7.7.1.0 2 ,?.0 1 3 , 1 ? heptadeca-1 16 ,2,4,6,9 17 ,10,12-heptaene-8,15-dione
PubChem CID 1080352
SMILES C1=CC=C(C=C1)CCNC2=C3C4=CC=CC=C4C(=O)C5=C3C(=CC=C5)NC2=O

Specifications

Quantity 50 mg
Formula Weight 366.41
Percent Purity >97%
Chemical Name or Material BRD 7389
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.