Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585662
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585662 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585662 Supplier Invitrogen™ Supplier No. PA585662
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Pig (98%). Positive Control: 293T nuclear extract. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

BRD4, a ubiquitously expressed nuclear phosphoprotein, is a member of BET protein family. It is a human homolog of mouse MCAP and consists of 2 N-terminal bromodomains, a central ET-domain, and several kinase-like motifs in its N-terminus. It is expressed as 2 isoforms; the longer variant has a C-terminus region with a praline and glutamine-rich domains. BRD4 is a chromatin-binding protein whose expression is induced in response to growth stimuli. BRD4 acts at different stages of the cell cycle by interacting with proteins including RFC and SPA-1 and regulates cell growth. It also has a role in regulating chromosomal dynamics during mitosis. BRD4 functions as a cellular target for E2 protein of papillomavirus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRD4
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene BRD4
Gene Accession No. O60885, Q9ESU6
Gene Alias Brd4; BRD4-NUT FUSION; Brd5; bromodomain containing 4; bromodomain-containing 4; bromodomain-containing 5; bromodomain-containing protein 4; bromodomain-containing protein 4; LOW QUALITY PROTEIN: bromodomain-containing protein 4; CAP; chromosome-associated protein; fsh-like protein; HUNK1; HUNKI; Mcap; mitotic chromosome-associated protein; Protein HUNK1; WI-11513
Gene Symbols BRD4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human BRD4 . The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23476, 57261
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.