Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRD8 / SMAP2 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993073
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3073 10 μL
50-199-3072 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3073 Supplier Active Motif Supplier No. 61008
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transcriptional repressor that contribute to growth arrest during terminal macrophage differentiation by repressing target genes involved in Ras-dependent proliferation. Represses MMP1 promoter activity.

Specifications

Antigen BRD8 / SMAP2
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene BRD8
Gene Accession No. Q9H0E9
Gene Alias 2610007E11Rik; 4432404P07Rik; BOS_8154; BRD8; bromodomain containing 8; Bromodomain-containing protein 8; p120; Skeletal muscle abundant protein; Skeletal muscle abundant protein 2; SMAP; SMAP2; Thyroid hormone receptor coactivating protein of 120 kDa; TrCP120
Gene Symbols BRD8
Host Species Rabbit
Immunogen This BRD8 / SMAP2 antibody was raised against a peptide corresponding to amino acids within the N-terminal region of human BRD8.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10902
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.