Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam BrdU Control Slides, Human MCF7 Cells, 5 Slides

Catalog No. 502655573
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 Slides
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-265-5573 5 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-265-5573 Supplier Abcam Supplier No. AB2813355SLIDE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Unstained, formalin-fixed, BrdU-labeled MCF7 cells on glass slides for IHC-P analysis.

  • Cell Type: MCF7 (human breast carcinoma derived) cells
  • Sample Type: Formalin-fixed, BrdU labeled cells on glass slides
  • Applications: IHC-P
  • Quantity: 5 slides

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule BrdU
Reactivity Human
Label or Dye BrdU
Format Glass Slides
Kit Contents 5 BrdU labeled MCF7 cell slides
Detection Location In-Cell Detection
Cross Reactivity Human
Sample Type Cells
Quantity 5 Slides
For Use With (Application) IHC-P
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.