Learn More
Description
Unconjugated MA3-071 has been successfully used in immunofluorescence, immunohistochemistry, and flow cytometry applications. DyLight 488 conjugated MA3-071-D488 has been successfully used in flow cytometry applications.
Specifications
Specifications
| Antigen | BrdU |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | BU-1 |
| Concentration | 1 mg/mL |
| Conjugate | DyLight 488 |
| Formulation | PBS with proprietary stabilizer and 0.02% sodium azide |
| Gene Alias | 5-Bromo-2-deoxyuridine; Bromodeoxyuridine |
| Host Species | Mouse |
| Immunogen | 5-iodouridine covalently coupled to ovalbumin. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
