Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BrdU Monoclonal Antibody (MoBU-1)
GREENER_CHOICE

Catalog No. B35128
Encompass_Preferred
Change view
Click to view available options
Quantity:
350 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35128 350 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B35128 Supplier Invitrogen™ Supplier No. B35128
  • $440.00
    $330.65 / Each of 1

    Save $109.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

The MoBU-1 clone has a high specificity for BrdU, in addition to no cross-reactivity with EdU. This clone can also readily detect bromouridine (BrU) incorporated into RNA or BrdUTP incorporated into DNA strand breaks via the TUNEL assay. Samples labeled with the Alexa Fluor™ conjugates of MoBU-1 can be directly imaged by fluorescence microscopy or high-content imaging. The unconjugated form and the biotin conjugate of this clone require a secondary antibody or a streptavidin conjugate for detection, respectively. The MoBU-1 clone is also available for flow cytometry applications.

Bromodeoxyuridine (5-bromo-2-deoxyuridine, BrdU) is a synthetic nucleoside that is an analogue of thymidine. BrdU is commonly used in the detection of proliferating cells in living tissues, and can be incorporated into the newly synthesized DNA of replicating cells (during the S phase of the cell cycle), substituting for thymidine during DNA replication. Antibodies specific for BrdU can then be used to detect the incorporated chemical, thus indicating cells that were actively replicating their DNA. Binding of the antibody requires denaturation of the DNA, usually by exposing the cells to acid or heat.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BrdU
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone MoBU-1
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 5mM sodium azide; pH 7.2
Gene Alias 5-Bromo-2-deoxyuridine; Bromodeoxyuridine
Host Species Mouse
Immunogen BrdU.
Purification Method Affinity chromatography
Quantity 350 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.