Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BrdU Monoclonal Antibody (MoBU-1), Alexa Fluor™ 647
GREENER_CHOICE

Catalog No. B35133
Encompass_Preferred
Change view
Click to view available options
Quantity:
350 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35133 350 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B35133 Supplier Invitrogen™ Supplier No. B35133
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Cell proliferation can be measured with BrdU following its incorporation into newly synthesized DNA and its subsequent detection with an anti-BrdU antibody. Although this method is rapidly being replaced with the recently developed click chemistry-based Click-iTTM EdU assay, an advantage of the antibody clone MoBU-1 is that it does not cross-react with the thymidine analog EdU (5-ethynyl-2'-deoxyuridine), thus making it valuable for use in dual-pulse applications. Traditionally, the dual-pulse method, which is used for studying biological events such as neurogenesis and cell differentiation or evaluating anti-cancer drug efficacy, employs BrdU immunocytochemistry with iododeoxyuridine (IdU) or chlorodeoxyuridine (CldU), using multiple antibodies for detection. However, using sequential pulses of the thymidine analogs EdU and BrdU greatly simplifies dual-pulse labeling, and allows a straightforward and reliable method of distinguishing BrdU- and EdU-labeled cells. In addition to no cross-reactivity with EdU, the MoBU-1 clone has a high specificity for BrdU. This clone can also readily detect bromouridine (BrU) incorporated into RNA or BrdUTP incorporated into DNA strand breaks via the TUNEL assay. Samples labeled with the Alexa FluorTM conjugates of MoBU-1 can be directly imaged by fluorescence microscopy or high-content imaging. The approximate fluorescence excitation/emission maxima for Alexa FluorTM647 conjugate are 650/665 or 670 in nm. Detection with BrdU antibodies re...

Bromodeoxyuridine (5-bromo-2-deoxyuridine, BrdU) is a synthetic nucleoside that is an analogue of thymidine. BrdU is commonly used in the detection of proliferating cells in living tissues, and can be incorporated into the newly synthesized DNA of replicating cells (during the S phase of the cell cycle), substituting for thymidine during DNA replication. Antibodies specific for BrdU can then be used to detect the incorporated chemical, thus indicating cells that were actively replicating their DNA. Binding of the antibody requires denaturation of the DNA, usually by exposing the cells to acid or heat.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BrdU
Applications Immunohistochemistry
Classification Monoclonal
Clone MoBU-1
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Formulation PBS with 5mM sodium azide; pH 7.2
Gene Alias 5-Bromo-2-deoxyuridine; Bromodeoxyuridine
Host Species Mouse
Immunogen BrdU.
Purification Method Affinity chromatography
Quantity 350 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.