Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BrdU Monoclonal Antibody (ZBU30)
GREENER_CHOICE

Catalog No. 033900
Encompass_Preferred
Change view
Click to view available options
Quantity:
400 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-390-0 400 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-390-0 Supplier Invitrogen™ Supplier No. 033900
  • $479.00
    $359.65 / Each of 1

    Save $119.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

BrdU Monoclonal Antibody for IHC

Bromodeoxyuridine (5-bromo-2-deoxyuridine, BrdU) is a synthetic nucleoside that is an analogue of thymidine. BrdU is commonly used in the detection of proliferating cells in living tissues, and can be incorporated into the newly synthesized DNA of replicating cells (during the S phase of the cell cycle), substituting for thymidine during DNA replication. Antibodies specific for BrdU can then be used to detect the incorporated chemical, thus indicating cells that were actively replicating their DNA. Binding of the antibody requires denaturation of the DNA, usually by exposing the cells to acid or heat.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BrdU
Applications Immunohistochemistry
Classification Monoclonal
Clone ZBU30
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Alias 5-Bromo-2-deoxyuridine; Bromodeoxyuridine
Host Species Mouse
Immunogen BrdU.
Purification Method Purified
Quantity 400 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Target Molecule Chemical
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.