Learn More
US Biomax Inc Breast cancer with cancer adjacent breast (match and unmatch) tissue array, 106 cases/110 cores (core size 1.5mm), replacing BC081116e
SDP
Supplier:  US Biomax Inc BC081116F
Encompass
Breast cancer with cancer adjacent breast (match and unmatch) tissue array, including pathology grade, TNM and clinical stage (AJCC 8th edition), IHC marker (ER/PR/HER2/Ki-67) result, 106 cases/110 cores (core size 1.5mm), replacing BC081116e
Catalog No. NC3237345
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.