Learn More
Amsbio LLC Breast cancer (including one case of male breast cancer) with adjacent normal breast tissue array
Supplier:  Amsbio LLC BC081120G
Encompass
Breast cancer with adjacent normal breast tissue array including pathology grade TNM and clinical stage with IHC results ER/PR/Her2 110 cases/110 cores replacing BC081120f
Catalog No. 50-256-4552