Learn More
R&D Systems™ Breast carcinoma amplified sequence 3 Antibody Blocking Peptide
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Gene ID (Entrez) | 54828 |
| Species | Human |
| Product Line | Novus |
| Content And Storage | Store at -20 C. Avoid freeze-thaw cycles. |
| Formulation | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-77125. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | BCAS4/BCAS3 fusion, breast carcinoma amplified sequence 3, breast carcinoma amplified sequence 4/3 fusion protein, breast carcinoma-amplified sequence 3, DKFZp686O1527, FLJ20128, GAOB1, MAAB, metastasis associated antigen of breast cancer, MGC4973, protein Maab1 |
| Gene Symbol | BCAS3 |
| Product Type | Breast carcinoma amplified sequence 3 |
| Quantity | 0.05 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.