Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DenLine™ Uniforms Protection Plus™ DL361 UltraLite™ Unisex Mid-Length Lab Jackets
SDP

Catalog No. 22412445
Change view
Click to view available options
Color:
Black
Ceil Blue
Royal Blue
White
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
28 product options available for selection
Product selection table with 28 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
22-412-445 Black X-Small
22-412-424 White X-Small
22-412-425 White Small
22-412-426 White Medium
22-412-427 White Large
22-412-428 White X-Large
22-412-429 White 2X-Large
22-412-430 White 3X-Large
22-412-431 Ceil Blue X-Small
22-412-432 Ceil Blue Small
22-412-433 Ceil Blue Medium
22-412-434 Ceil Blue Large
22-412-435 Ceil Blue X-Large
22-412-436 Ceil Blue 2X-Large
22-412-437 Ceil Blue 3X-Large
22-412-438 Royal Blue X-Small
22-412-439 Royal Blue Small
22-412-440 Royal Blue Medium
22-412-441 Royal Blue Large
22-412-442 Royal Blue X-Large
22-412-443 Royal Blue 2X-Large
22-412-444 Royal Blue 3X-Large
22-412-446 Black Small
22-412-447 Black Medium
22-412-448 Black Large
22-412-449 Black X-Large
22-412-450 Black 2X-Large
22-412-451 Black 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 28 options available.
28 options
This item is not returnable. View return policy
Catalog No. 22-412-445 Supplier DenLine™ Uniforms Supplier No. DL361BKXS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Breathable lab coats provide AAMI PB70 Level 2 fluid resistance and feature heavy duty stainless steel snap closures and hip side vents.

  • Primary Applications:
    • Anatomic Pathology
    • Clinical Pathology
    • Clinical Laboratory
    • Phlebotomy
    • Specimen Collection
    • Blood Collection, Testing
    • Manufacturing
    • Dental
    • Human and Animal Research
    • Cleanroom ISO 5/Class 100 - Medical Device, Pharma, Electronics
  • Complies with OSHA 29 CFR 1910
  • Provides 360 degree front and back, plus shoulder protection
  • AAMI PB70 Level 2
  • Overall Air Permeability 35 CFM (ASTM D737)
  • Stain resistant fabric and cuffs (blood, bleach and ALL aqueous reagents and liquids)
  • Anti-static fabric prevents false instrument readings, damage to computers
  • Low lint fabric prevents sample contamination, ideal for molecular testing
  • Durable construction, 3 pockets, double stitched
  • Stainless snap closures
  • Cuffs for overgloving
  • Bleach safe all colors
  • Hang loop, outside back of collar
  • Side vents for pant pocket access
  • Autoclavable fabric
  • 225 commercial wash tested

Specifications

Product Type General Purpose Lab Coat
Size X-Small
Gender Unisex
Material Polyester, Carbon
Color Black
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Static, Liquids, Stains
Style Mid-length
No. of Pockets 3
Collar Type Traditional
Latex Free Yes
Size (English) Chest 30 to 32 in.
Size (English) Hip 33 to 35 in.
Size (Metric) Chest 76 to 81 cm
Size (Metric) Hip 84 to 89 cm
Certifications/Compliance OSHA 29 CFR 1910, AAMI PB70 Level 2 Protection
For Use With (Application) Anatomic Pathology, Clinical Pathology, Clinical Laboratory, Phlebotomy, Specimen Collection, Research, Dental, Cleanrooms
Length (English) 34 in.
Length (English) Cuff 4 in.
Length (English) Sleeve 35 in.
Length (Metric) 86.3 cm
Length (Metric) Cuff 10.1 cm
Weight (English) 0.6 lb.
Autoclavable Autoclavable
Permeability Overall Air Permeability 35 CFM (ASTM D737)
Construction 360° Fluid Resistance Front, Shoulders and Back
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.