Learn More
Meridian Bioscience™ BreathID™ HP Smart
GSA_VA
The BreathID™ Smart is a urea breath test platform for the detection of H. pylori and confirmation of eradication.
Supplier:  Meridian Bioscience™ VS07000
Catalog No. 23-655-113
Request a Quote
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
The state-of-the-art BreathID™ Smart urea breath test platform for H. pylori detection intuitively combines market-leading diagnostic accuracy with efficient automation, all in a compact footprint for maximizing mid-range testing throughput. Whether for initial active infection detection or confirmation of eradication, Meridian is your trusted partner in gastrointestinal diagnostics.
  • Perfect platform for low to mid-volume laboratories and clinics
    • Test samples have a two-week window for analysis and require no special storage
    • 4 test samples per auto sampler
  • Under 10 minutes for full batch analysis, 2 minutes per test
  • User-friendly interface
    • Touchscreen for intuitive process management
    • Scannable barcodes maximize speed of data entry
    • Automated QC selection and label generation
  • Flexible data management options
    • Print, export, or transmit results
    • Secure, seamless integration with LIS
  • Minimizes potential errors before they happen
    • Automatic identification and correction of reversed bags
    • Coordinated indicator signals guide batch assembly and bag placement
    • Scannable barcodes minimize human error
Specifications
Specifications
Yes
H. pylori
4 Test samples per auto sampler
Breath Test System
97.90%
Waived
Kit
100%
Reusable
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.