Learn More
Enzo Life Sciences Brefeldin A (100mg). CAS: 20350-15-6
SDP
Supplier:  Enzo Life Sciences BMLG4050100
Encompass_Preferred
Inhibitor of intracellular protein transport and protein secretion, interfering with trafficking in the trans-Golgi network, leading to the accumulation of cycling proteins in ERGIC clusters. Blocks ADP-ribosylation factor (Arf) in an inactive GDP-bound conformation and thereby prevents binding of COPI coats to ERGIC and Golgi membranes. Upon brefeldin A treatment the Golgi rapidly tubulates and fuses with the ER by an energy-, temperature-, and microtubule-dependent process. In contrast, the drug has little effect on the ERGIC, which keeps its identity, although the ERGIC clusters become larger and more uniformly distributed in the cytoplasm of the cells. Inhibits intracellular collagen degradation. Also inhibits apical Na+ channels in epithelia. Induces apoptosis. Alternative name: BFA, Ascotoxin, Decumbin. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (20mg/ml), 100% ethanol (5mg/ml), methanol, acetone or ethyl acetate (1mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0879
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.