Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brefeldin A, >98%, MP Biomedicals™

Catalog No. ICN19480210 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19480210 10 mg
ICN19480205 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19480210 Supplier MP Biomedicals, Inc Supplier No. 0219480210
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20350-15-6
Molecular Formula C16H24O4
Molecular Weight (g/mol) 280.36
MDL Number MFCD00083258,MFCD12913297
InChI Key KQNZDYYTLMIZCT-PNFJWZTBSA-N
Synonym Ascotoxin, Cyanein
PubChem CID 133640201
IUPAC Name (1S,6S,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-1H,4H,6H,7H,8H,9H,11aH,12H,13H,14H,14aH-cyclopenta[f]oxacyclotridecan-4-one
SMILES C[C@H]1CCC\C=C\[C@@H]2C[C@H](O)C[C@H]2[C@@H](O)\C=C\C(=O)O1

Specifications

Type Brefeldin A
Physical Form Crystalline Powder
Color White
Quantity 10 mg
Formula Weight 280.37
Percent Purity >98%
Grade Not Available
Chemical Name or Material Brefeldin A
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.