CAS RN 20350-15-6
Brefeldin A, 98%
CAS: 20350-15-6 Molecular Formula: C16H24O4 Molecular Weight (g/mol): 280.36 MDL Number: MFCD00083258,MFCD12913297 InChI Key: KQNZDYYTLMIZCT-PNFJWZTBSA-N Synonym: brefeldin a,cyanein,decumbin,bfa PubChem CID: 133640201 SMILES: C[C@H]1CCC\C=C\[C@@H]2C[C@H](O)C[C@H]2[C@@H](O)\C=C\C(=O)O1
Brefeldin A, 99.4%, MP Biomedicals™
CAS: 20350-15-6 Molecular Formula: C16H24O4 Molecular Weight (g/mol): 280.36 MDL Number: MFCD00083258,MFCD12913297 InChI Key: KQNZDYYTLMIZCT-PNFJWZTBSA-N Synonym: brefeldin a,cyanein,decumbin,bfa PubChem CID: 133640201 IUPAC Name: (1S,6S,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-1H,4H,6H,7H,8H,9H,11aH,12H,13H,14H,14aH-cyclopenta[f]oxacyclotridecan-4-one SMILES: C[C@H]1CCC\C=C\[C@@H]2C[C@H](O)C[C@H]2[C@@H](O)\C=C\C(=O)O1
Brefeldin A, MP Biomedicals
CAS: 20350-15-6 Molecular Formula: C16H24O4 Molecular Weight (g/mol): 280.36 MDL Number: MFCD00083258,MFCD12913297 InChI Key: KQNZDYYTLMIZCT-PNFJWZTBSA-N PubChem CID: 133640201 IUPAC Name: (1S,6S,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-1H,4H,6H,7H,8H,9H,11aH,12H,13H,14H,14aH-cyclopenta[f]oxacyclotridecan-4-one SMILES: C[C@H]1CCC\C=C\[C@@H]2C[C@H](O)C[C@H]2[C@@H](O)\C=C\C(=O)O1
MilliporeSigma™ Calbiochem™ (+)-Brefeldin A, Eupenicillium brefeldianum
Specifically and reversibly blocks translocation of proteins from the endoplasmic reticulum (ER) to the Golgi apparatus without affecting endocytosis or lysosome function.
MilliporeSigma™ Calbiochem™ InSolution™ (+)-Brefeldin A, Eupenicillium brefeldianum
A 25 mM solution of (+)-Brefeldin A in DMSO.
Thermo Scientific Chemicals Brefeldin A, natural
CAS: 20350-15-6 Molecular Formula: C16H24O4 Molecular Weight (g/mol): 280.36 MDL Number: MFCD00083258,MFCD12913297 InChI Key: KQNZDYYTLMIZCT-PNFJWZTBSA-N Synonym: brefeldin a,cyanein,decumbin,bfa PubChem CID: 133640201 IUPAC Name: (1S,7S,10E,12R,13R,15S)-12,15-dihydroxy-7-methyl-8-oxabicyclo[11.3.0]hexadeca-2,10-dien-9-one SMILES: C[C@H]1CCC\C=C\[C@@H]2C[C@H](O)C[C@H]2[C@@H](O)\C=C\C(=O)O1
Brefeldin A, >98%, MP Biomedicals™
CAS: 20350-15-6 Molecular Formula: C16H24O4 Molecular Weight (g/mol): 280.36 MDL Number: MFCD00083258,MFCD12913297 InChI Key: KQNZDYYTLMIZCT-PNFJWZTBSA-N Synonym: Ascotoxin,Cyanein PubChem CID: 133640201 IUPAC Name: (1S,6S,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-1H,4H,6H,7H,8H,9H,11aH,12H,13H,14H,14aH-cyclopenta[f]oxacyclotridecan-4-one SMILES: C[C@H]1CCC\C=C\[C@@H]2C[C@H](O)C[C@H]2[C@@H](O)\C=C\C(=O)O1
R&D Systems™ Brefeldin A
CAS: 20350-15-6 Molecular Formula: C16H24O4