Learn More
Lkt Laboratories CAS 79580-28-2, ≥95%, Polyether neurotoxin found in Karenia brevis; Nav1.4/1.5 Na+ channel activator.
SDP
Supplier:  Lkt Laboratories B6917
Encompass_Preferred
Brevetoxins are lipid-soluble polyether neurotoxins initially produced by the dinoflagellate Karenia brevis. Brevetoxins act as agonists, binding the neurotoxin site 5 on SCN4A/Nav1.4 and SCN5A/Nav1.5 voltage-gated Na+ channels in skeletal muscle and cardiac tissue, producing a channel opening effect; this activity results in bronchoconstriction and airway inflammation in vivo. Stimulation of the voltage-gated Na+ channels also induces NMDA receptor-mediated release of L-glutamate and L-aspartate in neurons. Activity at these channels also activates Pyk2 and Src, inducing downstream signaling cascades. Brevetoxin 2 forms DNA adducts with cytidine in isolated lung cells and with adenosine and guanosine in vivo, indicating carcinogenic activity.
Catalog No. NC2522732
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.