Learn More
Lkt Laboratories Brevetoxin 3, ≥95%, 100UG
SDP
Supplier:  Lkt Laboratories B6918100UG
Encompass_Preferred
Polyether neurotoxin found in Karenia brevis; Nav1.4/1.5 Na+ channel activator. CAS: 85079-48-7. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. 50-153-4977
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.