Learn More
Description
Includes
7 test factors:
- Chloride - Direct reading titrator; 1 drop = 10, 25ppm; Approx 80 tests
- Sulfate - Turbidimetric; 0 to 200ppm sulfate; 50 tests
- Alkalinity - Direct reading titrator; 0 to 200ppm (and higher) as CaCO3; 50 test at 200ppm
- Total hardness - Direct reading titrator; 0 to 200ppm (and higher) as CaCO3; 50 tests at 200ppm
- Calcium hardness - Direct titrator reading; 0 to 200ppm (and higher) as CaCO3; 50 tests at 200ppm
- Magnesium hardness - Calculation
- Sodium - Calulation
The quality of the water that you start with will affect your final product, so take the mystery out of making great-tasting beer with the BrewLab™ Test Kit for home brewers.
- Economical, easy-to-use test kit to quickly measure the mineral content of water and allow the brewer to make informed decisions in the brewing process
- Alkalinity is the carbonate and bicarbonate content of the water, which acts to raise the pH of the mash and beer
- Chloride ion acts to bring out the sweetness and fullness of the malt flavor
- Dissolved oxygen can have both a positive and a negative impact on beer
- Fermentation itself is an anerobic process, but yeast cells require oxygen for growth
- Calcium helps prevent enzymes from denaturing and extends their activity in mash, promoting clarity, flavor and stability
- Magnesium is an important yeast nutrient
- The mineral content of the water directly affects the pH of the mash, and the pH of the mash affects the entire brewing process
- Sodium acts in concert with chloride to enhance the sweetness and fullness of the malt, but too much can oversalt the beer and result in salty or metallic flavors
- Sulfate ion acts to accenturate the hoppiness and dryness of the beer, making it more crisp tasting
- Measurement of total dissolved solids allows the brewer to quickly determine if there has been a shift or change in a water source or brewing process as well as provide a QA check on demineralization and waste treatment effectiveness
Specifications
Specifications
| For Use With (Application) | Quantifies 7 Water Quality Factors |
| Includes | Tests for 7 water quality factors |
| Product Type | BrewLab™ Basic |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

