Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFP36L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527445 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527445 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527445 Supplier Invitrogen™ Supplier No. PA527445
  • $622.50
    $498.65 / Each of 1

    Save $123.85

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (98%), Rat (98%), Chicken (89%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene is a member of the TIS11 family of early response genes. Family members are induced by various agonists such as the phorbol ester TPA and the polypeptide mitogen EGF. The gene is well conserved across species and has a promoter that contains motifs seen in other early-response genes. The encoded protein contains a distinguishing putative zinc finger domain with a repeating cys-his motif. This putative nuclear transcription factor most likely functions in regulating the response to growth factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFP36L1
Applications Western Blot
Classification Polyclonal
Concentration 6.34 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene ZFP36L1
Gene Accession No. Q07352
Gene Alias AW742437; AW743212; Berg36; Brf1; butyrate response factor 1; cMG1; D530020L18Rik; early response factor Berg36; EGF-inducible protein CMG1; EGF-response factor 1; ERF1; ERF-1; mRNA decay activator protein ZFP36L1; RNF162B; TIS11b; TPA-induced sequence 11b; ZFP36 ring finger protein like 1; ZFP36 ring finger protein-like 1; ZFP36L1; ZFP36-like 1; zinc finger protein 36, C3H type-like 1; zinc finger protein 36, C3H1 type-like 1; zinc finger protein, C3H type, 36-like 1
Gene Symbols ZFP36L1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TIS11B. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 677
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.