Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597155
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597155 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597155 Supplier Thermo Scientific Supplier No. PA597155
Only null left
Add to Cart
Add to Cart

BRF2 Polyclonal Antibody

BRF2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen BRF2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Brf2
Gene Accession No. Q3UAW9, Q4V8D6, Q9HAW0
Gene Alias 2700059M06Rik; 5730512K07Rik; AU018504; AU022602; B-related factor 2; BRF2; BRF-2; BRF2, RNA polymerase III transcription initiation factor 50 kDa subunit; BRF2, RNA polymerase III transcription initiation factor 50 subunit; BRF2, subunit of RNA polymerase III transcription initiation factor, BRF1-like; BRFU; FLJ11052; hBRFU; hTFIIIB50; PRO1470; RNA polymerase III transcription initiation factor BRFU; TFIIIB50; transcription factor IIB- related factor, TFIIIB50; transcription factor IIIB 50 kDa subunit
Gene Symbols Brf2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 180-419 of human BRF2 (NP_0607802).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 306542, 55290, 66653
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.