Learn More
Santa Cruz Biotechnology SMARCA4/Brg1 Antibody (H-10) X, sc-374197 X, 200 µg/0.1 ml
Supplier:  Santa Cruz Biotechnology SC374197 X
Encompass
MARCA4/Brg1 Antibody (H-10) is a mouse monoclonal IgG1 κ, cited in 15 publications, provided at 200 µg/mlspecific for an epitope mapping between amino acids 115-149 near the N-terminus of Brg-1 of human originAnti-SMARCA4/Brg1 Antibody (H-10) is recommended for detection of Brg-1 of mouse, rat and human origin by WB, IP, IF and ELISA; also reactive with additional species, including and canine, bovine and porcine
Catalog No. NC2868914
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.