Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BrightComp eBeads™ Compensation Bead Kits

Catalog No. A54741
Encompass_Preferred
Change view
Click to view available options
Description:
CFP BrightComp eBeads Compensation Bead Kit
GFP BrightComp eBeads Compensation Bead Kit
RFP BrightComp eBeads Compensation Bead Kit
YFP BrightComp eBeads Compensation Bead Kit
mCherry BrightComp eBeads Compensation Bead Kit
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
A54741 YFP BrightComp eBeads Compensation Bead Kit
A54740 RFP BrightComp eBeads Compensation Bead Kit
A54742 CFP BrightComp eBeads Compensation Bead Kit
A54743 mCherry BrightComp eBeads Compensation Bead Kit
A10514 GFP BrightComp eBeads Compensation Bead Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. A54741 Supplier Invitrogen™ Supplier No. A54741
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

BrightComp eBeads compensation beads provide a consistent, accurate, and simple-to-use technique for setting flow cytometry compensation when using fluorescent protein-expressing samples.

• Three levels of intensity, perfect for any samples no matter the level of expression
• Near-identical, spectrally-matched dye to fluorescent protein
• Consistent, reliable performance

Accurate compensation is crucial for effective multicolor analysis in flow cytometry. Each kit provides polystyrene microspheres with near-identical, spectrally-matched fluorescent protein dye as well as negative control beads in a single dropper bottle.
TRUSTED_SUSTAINABILITY

Specifications

Description YFP BrightComp eBeads Compensation Bead Kit
Quantity 25 tests
Excitation Wavelength Range 488 nm
For Use With (Application) Flow Cytometry
Product Type Compensation Beads
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.