Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brightness Buffer, Reagents

Catalog No. 01335808
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-808 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-808 Supplier Reagents Holdings Llc Supplier No. CB10190020F
Only null left
Add to Cart
Add to Cart

Brightness Buffer, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Buffer
CAS 7732-18-5
Color Colorless
Physical Form Liquid
Quantity 20 L
Packaging Cubitainer
Sterility Non-sterile
Solubility Information Completely miscible
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.