Learn More
Alkali Scientific BrightStar™ PVDF Membrane, Pore Size 0.45µm, 300 X 3000 mm, 1 Roll
SDP
Supplier:  Alkali Scientific XR732
Encompass_Preferred
BrightStar™ 0.45µm Pore Size PVDF Membrane is perfect for Western, Northern, and Southern blotting of low molecular weight proteins and nucleic acids. It wets readily with water-based solutions and is easy to handle. Compatible with standard immunoblotting and nucleic acid detection methods. BrightStar™ Nitrocellulose Membrane offers superior binding capacity with no background interference. One roll - 300 x 3000mm (30cm wide x 3.0m length).
Catalog No. 50-233-5960
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.